missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane, 95%

Product Code. 11399275
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399275 25 g 25g
11389275 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399275 Supplier Thermo Scientific Alfa Aesar Supplier No. L16604.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102488-47-1 | C10H10ClF13Si | 440.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102488-47-1
Molecular Formula C10H10ClF13Si
Molecular Weight (g/mol) 440.70
MDL Number MFCD00042359
InChI Key AKYGPHVLITVSJE-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorooctyldimethylchlorosilane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl dimethylchlorosilane, 2-tridecafluorohexyl ethyldimethylchlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-dimethylchlorosilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyldimethylchlorosilane, chloro-dimethyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-n-octyl silane, silane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl
PubChem CID 145382
IUPAC Name chloro-dimethyl-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES C[Si](C)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Boiling Point 189°C to 191°C
Quantity 25 g
Flash Point 52°C (125°F)
Refractive Index 1.3453
Sensitivity Moisture sensitive
Density 1.461
UN Number UN2986
Beilstein 7040835
Formula Weight 440.71
Percent Purity 95%
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.