missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorodecyltrichlorosilane, 96%, Thermo Scientific™

Product Code. p-7058455
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11399245

Brand: Thermo Scientific Alfa Aesar L16584.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H,2H-Perfluorodecyltrichlorosilane is a halogenated compound used for biochemical research. It is applied to movable microparts of microelectromechanical systems , thereby reducing the surface energy and preventing the stickiness. Further, it is used to coat micro- and nano-features on stamps for a nanoimprint lithography. In addition, it is used in the preparation of (1H,1H,2H,2H-heptadecafluorodecyl)trimethoxysilane by reacting with methanol.

Solubility
Miscible with tetrahydrofuran, tetrhydropyran, toluene and other organic solvents.

Notes
Moisture sensitive. Incompatible with oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78560-44-8
Molecular Formula C10H4Cl3F17Si
Molecular Weight (g/mol) 581.55
MDL Number MFCD00042344
InChI Key VIFIHLXNOOCGLJ-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorodecyltrichlorosilane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl, 1h,1h,2h,2h-perfluorotrichlorosilane, trichloro 1h,1h,2h,2h-perfluorodecyl silane, trichloro 1h,1h,2h,2h-heptadecafluorodecyl silane, 1-trichlorosilyl-1h,1h,2h,2h-heptadecafluorodecane, 1h,1h,2h,2h-perfluorodecyltrichlorosilane 1g, heptadecafluoro-1,1,2,2-tetrahydrodecyl trichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-trichlorosilane
PubChem CID 123577
IUPAC Name trichloro(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 224°C
Physical Form Liquid
Quantity 1 g
Linear Formula CF3(CF2)7(CH2)2SiCl3
UN Number UN2987
Beilstein 5889653
Flash Point >200°C (392°F)
Solubility Information Miscible with tetrahydrofuran,tetrhydropyran,toluene and other organic solvents.
Formula Weight 581.56
Percent Purity 96%
Refractive Index 1.348
Sensitivity Moisture sensitive
Density 1.54
Chemical Name or Material 1H,1H,2H,2H-Perfluorodecyltrichlorosilane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.