missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorodecyldimethylchlorosilane, tech. 90%, Thermo Scientific™

Product Code. 11339245
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11339245

Brand: Thermo Scientific Alfa Aesar L16582.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74612-30-9
Molecular Formula C12H10ClF17Si
Molecular Weight (g/mol) 540.72
MDL Number MFCD00042343
InChI Key JHCJWHBMXWOYDE-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorodecyldimethylchlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl dimethylchlorosilane, 2-heptadecafluorooctyl ethylchlorodimethylsilane, chloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dimethylsilane, chloro-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl-dimethylsilane, chlorodimethyl-3,3,4,4,5,5,6,6,7,7,8,8, 9,9,10,10,10-heptadecafluorodecylsilane, silane,chloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl dimethyl
PubChem CID 144665
IUPAC Name chloro-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)-dimethylsilane
SMILES C[Si](C)(CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)Cl

Specifications

Boiling Point 197°C to 198°C
Quantity 1 g
UN Number UN2987
Beilstein 4584072
Formula Weight 540.73
Percent Purity 90%
Grade Technical
Chemical Name or Material 1H,1H,2H,2H-Perfluorodecyldimethylchlorosilane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.