missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluoro-1-iodooctane, 97%, stab. with copper

Product Code. 11395339
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395339 5 g 5g
11305349 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395339 Supplier Thermo Scientific Alfa Aesar Supplier No. B20984.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2043-57-4 | C8H4F13I | 474.004 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are important intermediates in the synthesis of organic fluoride products. It has also been shown to have potential estrogenic effects.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from alkali metals, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2043-57-4
Molecular Formula C8H4F13I
Molecular Weight (g/mol) 474.004
MDL Number MFCD00039410
InChI Key NVVZEKTVIXIUKW-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorooctyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodo, 1-iodo-1h,1h,2h,2h-perfluorooctane, 1,1,2,2-tetrahydroperfluorooctyliodide, 1,1,2,2-tetrahydroperfluorooctyl iodide, 2-perfluorohexyl ethyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-iodooctane, 1h,1h,2h,2h-tridecafluoro-n-octyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl iodide, acmc-1ccsn
PubChem CID 74888
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodooctane
SMILES C(CI)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point ∼20°C
Density 1.94
Boiling Point 178°C to 180°C
Flash Point 150°C (302°F)
Refractive Index 1.36
Quantity 5 g
Beilstein 1887105
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 474
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-iodooctane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.