missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,2H,2H-Perfluoro-1-iododecane, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used in that preparation of a chemical compounds employed in high-performance organic thin-film transistors. It also finds it application as an appliance in boat and building paint.
Solubility
Insoluble in water.
Notes
Light sensitive. Store away from alkali metals bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used in that preparation of a chemical compounds employed in high-performance organic thin-film transistors. It also finds it application as an appliance in boat and building paint.
Solubility
Insoluble in water.
Notes
Light sensitive. Store away from alkali metals bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 53°C to 57°C |
| Density | 1.88 |
| Boiling Point | 226°C |
| Flash Point | None |
| Linear Formula | CF3(CF2)7CH2CH2I |
| Quantity | 5 g |
| Beilstein | 1894667 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 574.02 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?