missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluoro-1-iododecane, 97%, Thermo Scientific™

Product Code. 11315339
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11315339 5 g 5g
11325339 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11315339 Supplier Thermo Scientific Alfa Aesar Supplier No. B20894.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in that preparation of a chemical compounds employed in high-performance organic thin-film transistors. It also finds it application as an appliance in boat and building paint.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from alkali metals bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2043-53-0
Molecular Formula C10H4F17I
Molecular Weight (g/mol) 574.02
MDL Number MFCD00039411
InChI Key XVKJSLBVVRCOIT-UHFFFAOYSA-N
Synonym 2-perfluorooctyl ethyl iodide, 1-iodo-1h,1h,2h,2h-perfluorodecane, 1h,1h,2h,2h-perfluorodecyliodide, 1h,1h,2h,2h-perfluorodecyl iodide, decane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iodo, 1h,1h,2h,2h-perfluoro-1-iododecane, 1h,1h,2h,2h-heptadecafluorodecyl iodide, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iodo-decane, acmc-20ap95, c8f17ch2ch2i
PubChem CID 74885
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-10-iododecane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCI

Specifications

Melting Point 53°C to 57°C
Density 1.88
Boiling Point 226°C
Flash Point None
Linear Formula CF3(CF2)7CH2CH2I
Quantity 5 g
Beilstein 1894667
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 574.02
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-iododecane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.