missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized

Product Code. 10545671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10545671 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10545671 Supplier Thermo Scientific Acros Supplier No. 348610050
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 27905-45-9 | C13H7F17O2 | 518.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 27905-45-9
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 100.0°C (4.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C13H7F17O2
Refractive Index 1.3360 to 1.3380
MDL Number MFCD00042306
Quantity 5 g
Synonym 1h,1h,2h,2h-heptadecafluorodecyl acrylate, 1h,1h,2h,2h-perfluorodecyl acrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl acrylate, unii-y9bfb4689z, 2-perfluorooctyl ethyl acrylate, 1,1,2,2-tetrahydroperfluorodecyl acrylate, perfluorooctyl ethyl acrylate, 2-propenoic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester, 2-perfluorooctyl ethylacrylate, c8f17ch2ch2ococh=ch2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, benzene, hexane
InChI Key QUKRIOLKOHUUBM-UHFFFAOYSA-N
SMILES C=CC(=O)OCCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl prop-2-enoate
Stabilizer 90 to 110ppm MEHQ
Molecular Weight (g/mol) 518.17
PubChem CID 119747
Viscosity 12 mPa.s (25°C)
Formula Weight 518.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1H,1H,2H,2H-Heptadecafluorodecyl acrylate, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.