missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,11H-Perfluoroundecyl acrylate, tech. 85%, stab. with ca 50ppm 4-methoxyphenol

Product Code. 11369195
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11369195

Brand: Thermo Scientific Alfa Aesar L16488.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4998-38-3 | C14H6F20O2 | 586.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4998-38-3
Molecular Formula C14H6F20O2
Molecular Weight (g/mol) 586.17
MDL Number MFCD00042112
InChI Key HBFWXGZKVGSZKO-UHFFFAOYNA-N
Synonym 1h,1h,11h-eicosafluoroundecyl acrylate, 1h,1h,11h-perfluoroundecyl acrylate, acrylic acid 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecyl ester
PubChem CID 2776419
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecyl prop-2-enoate
SMILES FCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)OC(=O)C=C

Specifications

Melting Point 44°C to 53°C
Boiling Point 115°C (1 mmHg)
Quantity 25 g
Formula Weight 586.17
Percent Purity 85%
Grade Technical
Chemical Name or Material 1H,1H,11H-Perfluoroundecyl acrylate, stabilized with ≈50ppm 4-methoxyphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.