missing translation for 'onlineSavingsMsg'
Learn More

1H-1,2,4-Triazole-1-carboxamidine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15425786
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19503-26-5
Molecular Formula C3H6ClN5
Molecular Weight (g/mol) 147.566
MDL Number MFCD03095468
InChI Key JDDXNENZFOOLTP-UHFFFAOYSA-N
Synonym 1h-1,2,4-triazole-1-carboximidamide hydrochloride, 1h-1,2,4-triazole-1-carboxamidine monohydrochloride, 1h-1,2,4-triazole-1-carboxamidine hydrochloride, 1,2,4-triazole-1-carboximidamide hydrochloride, 1h-1,2,4-triazole-1-carboximidamide, monohydrochloride, c3h5n5.clh, ksc496s5d, 1,2,4-triazolecarboxamidine, chloride, 1-amidino-1,2,4-triazole hydrochloride, 1-amidino-1h-1,2,4-triazole hydrochloride
PubChem CID 16218515
IUPAC Name 1,2,4-triazole-1-carboximidamide;hydrochloride
SMILES C1=NN(C=N1)C(=N)N.Cl

Specifications

Melting Point 215°C to 220°C
Quantity 5 g
Beilstein 9500271
Formula Weight 147.57
Percent Purity 98%
Chemical Name or Material 1H-1,2,4-Triazole-1-carboxamidine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'