missing translation for 'onlineSavingsMsg'
Learn More

1H-1,2,3-Triazole, 98%

Product Code. p-7023440
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438846 1 g 1g
11448846 5 g 5g
11458846 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438846 Supplier Thermo Scientific Alfa Aesar Supplier No. A16999.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 288-36-8 | C2H3N3 | 69.067 g/mol

It effectively promotes the proton conduction in polymer electrolyte membranes via an intermolecular proton-transfer mechanism. A useful triazole for proteomics research

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It effectively promotes the proton conduction in polymer electrolyte membranes via an intermolecular proton-transfer mechanism. A useful triazole for proteomics research

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-36-8
Molecular Formula C2H3N3
Molecular Weight (g/mol) 69.067
MDL Number MFCD00014490
InChI Key QWENRTYMTSOGBR-UHFFFAOYSA-N
Synonym 1h-1,2,3-triazole, 1,2,3-triazole, 2h-1,2,3-triazole, 1,2,3-1h-triazole, triazole, triazol, osotriazole, v-triazole, 1,2,3-triazol, 1,2,3 triazole
PubChem CID 67516
ChEBI CHEBI:35565
IUPAC Name 2H-triazole
SMILES C1=NNN=C1

Specifications

Melting Point 23°C to 25°C
Density 1.192
Boiling Point 208°C
Flash Point 107°C (224°F)
Refractive Index 1.498
Quantity 1 g
Beilstein 104766
Solubility Information Fully miscible in water.
Formula Weight 69.07
Percent Purity 98%
Chemical Name or Material 1H-1,2,3-Triazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.