missing translation for 'onlineSavingsMsg'
Learn More

Testosterone, TRC

Product Code. 30424074
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30424074

Brand: Toronto Research Chemicals TRCT1550001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone
CAS 58-22-0
Purity Grade Notes HPLC
Molecular Formula C19 H28 O2
Quantity 1 g
Synonym 17beta-Hydroxyandrost-4-en-3-one, Testosterone, Testosterone Enantate Imp. D (EP), Testosterone Propionate Imp. C (EP), Testosterone Isocaproate Imp. A (EP), Androst-4-en-3-one','17-hydroxy-','(17β)-, Testosterone (7CI,8CI), (17β)-17-Hydroxyandrost-4-en-3-one, (+)-Testosterone, 17β-Hydroxy-Δ4-androsten-3-one, 17β-Hydroxyandrost-4-en-3-one, 17β-Hydroxyandrost-4-ene-3-one, 17β-Testosterone, 4-Androsten-3-one-17β-ol, AA 2500, Andro 100, AndroForte 5, AndroGel, Androderm, Androlin, Andronaq, Andropatch, Androst-4-en-17β-ol-3-one, Androst-4-ene-17β-ol-3-one, Andrusol, Axiron, CDB 111C, COL 1621, CP 601B, Compleo, Cristerona T, Geno-cristaux Gremy, Glowmin, Homosteron, Homosterone, Intrinsa, Mertestate, NSC 9700, Nasobol, Neotestis, Noseafix, Opterone, Oreton, Orquisteron, Perandren, Percutacrine androgenique, Primotest, Primoteston, Relibra, Striant, Synandrol F, Teslen, Testandrone, Testiculosterone, Testim, Testobase, Testoderm, Testogel, Testolent, Testolin, Testopatch, Testopel, Testopropon, Testosteroid, Testosteron, Testoviron Schering, Testoviron T, Testro AQ, Testrone, Testryl, Tostrelle, Tostrex, Viatrel, Virormone, Virosterone, Δ4-Androsten-17β-ol-3-one
InChI Formula InChI=1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O
IUPAC Name (8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 288.42
Formula Weight 288.2089
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.