Learn More
Ebastine, Thermo Scientific™
A histamine H1 receptor antagonist
Brand: Thermo Scientific Alfa Aesar J63088.03
| Quantity | 1g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySlightly soluble in chloroform, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
| 90729-43-4 | |
| MFCD00865661 | |
| 14,3484 | |
| Slightly soluble in chloroform,methanol. | |
| CC(C)(C)C1=CC=C(C=C1)C(=O)CCCN2CCC(CC2)OC(C3=CC=CC=C3)C4=CC=CC=C4 | |
| 469.669 | |
| 469.66 |
| C32H39NO2 | |
| 1g | |
| ebastine, kestine, ebastel, ebastin, kestin, evastel, bactil, ebastinum latin, las w-090, ebastina spanish | |
| MJJALKDDGIKVBE-UHFFFAOYSA-N | |
| 4-(4-benzhydryloxypiperidin-1-yl)-1-(4-tert-butylphenyl)butan-1-one | |
| 3191 | |
| Ebastine |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.