missing translation for 'onlineSavingsMsg'
Learn More

Buclizine, Dihydrochloride, TRC

Product Code. 30007609
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007609 1 g 1g
30132692 50 mg 50mg
30137516 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007609 Supplier Toronto Research Chemicals Supplier No. TRCB6894501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-74-8
Molecular Formula C28 H33 Cl N2 . 2 Cl H
Molecular Weight (g/mol) 505.95
Synonym Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-[[4-(1,1-dimethylethyl)phenyl]methyl]-, hydrochloride (1:2), Piperazine, 1-(p-tert-butylbenzyl)-4-(p-chloro-α-phenylbenzyl)-, dihydrochloride (6CI,7CI,8CI), Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-[[4-(1,1-dimethylethyl)phenyl]methyl]-, dihydrochloride (9CI), 1-(p-Chlorobenzhydryl)-4-(p-tert-butylbenzyl)diethylenediamine hydrochloride, 1-(p-tert-Butylbenzyl)-4-(p-chlorodiphenylmethyl)piperazine dihydrochloride, 1-p-Chlorobenzhydryl-4-p-(t)-butylbenzylpiperazine dihydrochloride, Aphilan R, Bucladin S, Buclina, Buclizine dihydrochloride, Buclizine hydrochloride, Buclodin, Histabutyzine hydrochloride, Longifene, NSC 25141, Softran, UCB 4445, Vibazine, Vibazine Hydrochloride
IUPAC Name 1-[(4-tert-butylphenyl)methyl]-4-[(4-chlorophenyl)-phenylmethyl]piperazine;dihydrochloride
SMILES Cl.Cl.CC(C)(C)c1ccc(CN2CCN(CC2)C(c3ccccc3)c4ccc(Cl)cc4)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C28H33ClN2.2ClH/c1-28(2,3)25-13-9-22(10-14-25)21-30-17-19-31(20-18-30)27(23-7-5-4-6-8-23)24-11-15-26(29)16-12-24;;/h4-16,27H,17-21H2,1-3H3;2*1H
Formula Weight 504.1866
Chemical Name or Material Buclizine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.