missing translation for 'onlineSavingsMsg'
Learn More

1Beta-Hydroxy Vitamin D3, TRC

Product Code. 30401128
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401128 10 mg -
30392534 25 mg 25mg
30405791 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30401128 Supplier Toronto Research Chemicals Supplier No. TRCH99583010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63181-13-5
Molecular Formula C27H44O2
Molecular Weight (g/mol) 400.64
Synonym (1beta,3beta,5Z,7E)- 9,10-Secocholesta-5,7,10(19)-triene-1,3-diol, 1beta-Calcidol, Alfacalcidol Impurity B (EP), (5Z,7E)-9,10-secocholesta-5,7,10(19)-triene-1beta,3beta-diol
IUPAC Name (1R,3R,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)C[C@@H](O)C3=C)\CCC[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H44O2/c1-18(2)8-6-9-19(3)24-13-14-25-21(10-7-15-27(24,25)5)11-12-22-16-23(28)17-26(29)20(22)4/h11-12,18-19,23-26,28-29H,4,6-10,13-17H2,1-3,5H3/b21-11+,22-12-/t19-,23-,24-,25+,26-,27-/m1/s1
Formula Weight 400.3341
Chemical Name or Material 1beta-Hydroxy Vitamin D3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.