missing translation for 'onlineSavingsMsg'
Learn More

1A-116, MedChemExpress

Product Code. 16474966
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16474966

Brand: Medchem Express HY104064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 1A-116, a potent Rac1 inhibitor, is specific for W56 residues, can prevent EGF-induced Rac1 activation and block Rac1-P-Rex1 interaction. 1A-116 can induce apoptosis and inhibit cell proliferation, migration and cycle progression in a concentration-dependent manner. 1A-116 also demonstrates a high antimetastatic activity in vivo.

1A-116, a potent Rac1 inhibitor, is specific for W56 residues, can prevent EGF-induced Rac1 activation and block Rac1-P-Rex1 interaction. 1A-116 can induce apoptosis and inhibit cell proliferation, migration and cycle progression in a concentration-dependent manner. 1A-116 also demonstrates a high antimetastatic activity in vivo.

Specifications

Chemical Name or Material 1A-116
Physical Form Solid
CAS 1430208-73-3
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C16H16F3N3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 100 mg/mL (325.40 mM; Need ultrasonic)
SMILES N=C(NC1=CC=CC=C1C(F)(F)F)NC2=CC(C)=CC(C)=C2
Molecular Weight (g/mol) 307.31
Formula Weight 307.31
Percent Purity 99.39%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.