missing translation for 'onlineSavingsMsg'
Learn More

18-Pentatriacontanone, tech. 85%, Thermo Scientific Chemicals

Catalog Number p-7051526
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
504-53-0
C35H70O
506.94
MFCD00048491
DMCJFWXGXUEHFD-UHFFFAOYSA-N
18-pentatriacontanone, stearone, heptadecyl ketone, diheptadecyl ketone, di-n-heptadecyl ketone, unii-7in8ga6m68, stearon, stearyl ketone, di-stearyl ketone, acmc-1asrl
10440
pentatriacontan-18-one
CCCCCCCCCCCCCCCCCC(=O)CCCCCCCCCCCCCCCCC
This item is not returnable. View return policy
504-53-0
C35H70O
506.94
MFCD00048491
DMCJFWXGXUEHFD-UHFFFAOYSA-N
18-pentatriacontanone, stearone, heptadecyl ketone, diheptadecyl ketone, di-n-heptadecyl ketone, unii-7in8ga6m68, stearon, stearyl ketone, di-stearyl ketone, acmc-1asrl
10440
pentatriacontan-18-one
CCCCCCCCCCCCCCCCCC(=O)CCCCCCCCCCCCCCCCC

Antiblocking agent

18-Pentatriacontanone is used as ant blocking agent. It is also used as pharmaceutical intermediate and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 79°C to 80°C
Color White
Linear Formula (CH3(CH2)16)2CO
Quantity 25 g
Solubility Information Soluble in benzene
Formula Weight 506.94
Percent Purity 85%
Physical Form Powder
Chemical Name or Material 18-Pentatriacontanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'