missing translation for 'onlineSavingsMsg'
Learn More

18-Hydroxyprogesterone, TRC

Product Code. 30390352
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390352 50 mg -
30407430 10 mg 10mg
30394362 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390352 Supplier Toronto Research Chemicals Supplier No. TRCH95066050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-69-0
Molecular Formula C21H30O3
Molecular Weight (g/mol) 330.46
Synonym 18-Hydroxypregn-4-ene-3,20-dione
IUPAC Name (8R,9S,10R,13R,14S,17S)-17-acetyl-13-(hydroxymethyl)-10-methyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12CO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H30O3/c1-13(23)17-5-6-19-16-4-3-14-11-15(24)7-9-20(14,2)18(16)8-10-21(17,19)12-22/h11,16-19,22H,3-10,12H2,1-2H3/t16-,17-,18+,19+,20+,21+/m1/s1
Formula Weight 330.2195
Chemical Name or Material 18-Hydroxyprogesterone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.