missing translation for 'onlineSavingsMsg'
Learn More

18β-Glycyrrhetinic acid, MedChemExpress

Product Code. 16451328
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451328 5 g 5g
16402366 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451328 Supplier Medchem Express Supplier No. HYN0180
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 18β-Glycyrrhetinic acid is the major bioactive component of Glycyrrhizae Radix and possesses anti-ulcerative, anti-inflammatory and antiproliferative properties.

18β-Glycyrrhetinic acid is the major bioactive component of Glycyrrhizae Radix and possesses anti-ulcerative, anti-inflammatory and antiproliferative properties.

Specifications

Chemical Name or Material 18β-Glycyrrhetinic acid
CAS 471-53-4
Color White
Purity Grade Notes Research
Molecular Formula C30H46O4
Quantity 5 g
Solubility Information DMSO : ≥ 250 mg/mL (531.15 mM)
SMILES CC1(C)[C@@H](O)CC[C@]([C@@]1([H])CC[C@@]([C@@]2(CC[C@]3(CC[C@](C(O)=O)(C[C@]3(C2=C4)[H])C)C)C)5C)(C)[C@@]5([H])C4=O
Molecular Weight (g/mol) 470.68
Formula Weight 470.68
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.