Learn More
Spare Pyrex™ stopcock glass keys
Click to view available options
Material:
Glass
Unit Size:
Each
Description
- Replacement keys for SSD: series stopcocks
- Manufactured from borosilicate glass
- Supplied complete with key retaining device
Specifications
Specifications
| Material | Glass |
| Product Type | Glass key and retaining set |