missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Retort Rings

Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Retort Rings comes in three types.

Brand:  Bochem™ 5500

Code : A0

Additional Details : Weight : 0.12330kg

 View more versions of this product

Product Code. 10578842

  • £23.40 / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

  • Made from 18/10 stainless steel
  • Three types: Closed ring, Closed ring with bosshead, and Open ring
Specifications

Specifications

Stainless Steel (18/10)
50 mm
1 Pk.
160 mm
Closed Ring
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers