missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Retort Rings

Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Retort Rings comes in three types.

Brand:  Bochem™ 5503

Code : A0

Additional Details : Weight : 0.29880kg

 View more versions of this product

Product Code. 10528711

  • £33.95 / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

  • Made from 18/10 stainless steel
  • Three types: Closed ring, Closed ring with bosshead, and Open ring
Specifications

Specifications

Stainless Steel (18/10)
120 mm
1 Pk.
220 mm
Closed Ring
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers