missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel One-Sided Chemical Spoons

Product Code. 10146500
Change view
Click to view available options
Length (Metric):
120 mm
150 mm
180 mm
210 mm
300 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10146500 120 mm Each
10409870 180 mm Each
10330581 150 mm Each
10588842 210 mm Each
10598842 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10146500 Supplier Bochem™ Supplier No. 3450
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from 18/10 stainless steel

  • One-sided chemical spoon
  • 18/10 stainless steel

Specifications

Quantity 1 Ea.
Product Type Chemical Scoop
Diameter (Metric) Outer 5mm
Handle Material Stainless Steel (18/10)
Material Stainless Steel (18/10)
Length (Metric) 120 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.