missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Evaporating Dish

Brand:  Bochem™ 8560

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10109550

  • £24.15 / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Flat
Stainless Steel
Evaporating Dish
75 mm
125 mL
Round
35 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers