Learn More
Bochem™ 18/10 Stainless Steel Cross Type Boss heads
Certified DIN 12895. Bochem™ 18/10 Stainless Steel Cross Type Boss heads comes in three types.
Brand:  Bochem™ 5306
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10350581
  • £81.65 / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Description
Description
  • 18/10 stainless steel
  • DIN 12895
  • Three types: Heart screw M6; Socket screw M6; and safety screw type "heart" M8
Specifications
Specifications
Stainless Steel (18/10)
Cross Type Boss head
8 mm
1 Pk.