missing translation for 'onlineSavingsMsg'
Learn More

17beta-Hydroxyprogesterone, TRC

Product Code. 30392926
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392926 100 mg
30406342 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392926 Supplier Toronto Research Chemicals Supplier No. TRCH950195100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-09-1
Molecular Formula C21H30O3
Molecular Weight (g/mol) 330.46
Synonym 17-Hydroxyisopregn-4-ene-3,20-dione, (17α)-17-Hydroxypregn-4-ene-3,20-dione
IUPAC Name (8R,10R,13R,14S,17S)-17-acetyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES O=C1CC[C@@]2(C)C(CC[C@]3([H])C2([H])CC[C@]4(C)[C@@]3([H])CC[C@]4(C(C)=O)O)=C1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H30O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h12,16-18,24H,4-11H2,1-3H3/t16-,17?,18+,19+,20-,21-/m1/s1
Formula Weight 330.2195
Chemical Name or Material 17beta-Hydroxyprogesterone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.