missing translation for 'onlineSavingsMsg'
Learn More

17Beta-Hydroxy Exemestane-d3, TRC

Product Code. 30392955
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392955 1 mg
30389832 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392955 Supplier Toronto Research Chemicals Supplier No. TRCH9423421MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C20H23D3O2
Molecular Weight (g/mol) 301.44
Synonym (17β)-17-Hydroxy-6-methyleneandrosta-1,4-dien-3-one-d3, Methylene Boldenone-d3
SMILES O=C1C=C[C@@]2(C)C(C(C[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC([2H])([2H])[C@]4([2H])O)=C)=C1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C26H34O8/c1-12-10-14-15-4-5-18(33-24-21(30)19(28)20(29)22(34-24)23(31)32)26(15,3)9-7-16(14)25(2)8-6-13(27)11-17(12)25/h6,8,11,14-16,18-22,24,28-30H,1,4-5,7,9-10H2,2-3H3,(H,31,32)/t14-,15-,16-,18-,19-,20-,21+,22-,24+,25+,26-/m0/s1/i5D2,18D
Chemical Name or Material 17beta-Hydroxy Exemestane-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.