missing translation for 'onlineSavingsMsg'
Learn More

17beta-Estradiol 3-O-Sulfate Sodium, TRC

Product Code. 30382111
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382111 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382111 Supplier Toronto Research Chemicals Supplier No. TRCE8888251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17beta-Estradiol 3-O-Sulfate Sodium
CAS 4999-79-5
Purity Grade Notes HPLC
Molecular Formula C18 H23 O5 S . Na
Quantity 1 g
Synonym Estra-1,3,5(10)-triene-3,17-diol (17β)-, 3-(hydrogen sulfate), sodium salt (1:1), Estra-1,3,5(10)-triene-3,17-diol (17β)-, 3-(hydrogen sulfate), monosodium salt (9CI), Estradiol, 3-(hydrogen sulfate), monosodium salt (6CI,7CI,8CI), 17β-Estradiol sodium sulfate
InChI Formula InChI=1S/C18H24O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3,5,10,14-17,19H,2,4,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,15-,16+,17+,18+;/m1./s1
SMILES [Na+].C[C@]12CC[C@H]3[C@@H](CCc4cc(OS(=O)(=O)[O-])ccc34)[C@@H]1CC[C@@H]2O
IUPAC Name sodium;[(8R,9S,13S,14S,17S)-17-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] sulfate
Molecular Weight (g/mol) 374.43
Formula Weight 374.1164
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.