missing translation for 'onlineSavingsMsg'
Få mere at vide

(17Beta)-3,3-Dimethoxyestra-5(10),9(11)-dien-17-ol-d3, TRC

Artikelnummer. 30377743
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
5 mg
10 mg
25 mg
Pakningsstørrelse:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30377743 10 mg 10mg
30378981 5 mg 5mg
30380236 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377743 Leverandør Toronto Research Chemicals Leverandørnr. TRCD89758710MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material (17beta)-3,3-Dimethoxyestra-5(10),9(11)-dien-17-ol-d3
Molecular Formula C20H27D3O3
Quantity 10 mg
Synonym 17β-Hydroxyestra-5(10),9(11)-dien-3-one Dimethyl Acetal-d3
InChI Formula InChI=1S/C20H30O3/c1-19-10-8-15-14-9-11-20(22-2,23-3)12-13(14)4-5-16(15)17(19)6-7-18(19)21/h8,16-18,21H,4-7,9-12H2,1-3H3/t16-,17+,18+,19+/m1/s1/i7D2,18D
SMILES O[C@@]1([2H])C([2H])([2H])C[C@@]2([H])[C@](C3=CC[C@@]21C)([H])CCC4=C3CCC(OC)(OC)C4
IUPAC Name (8S,13S,14S,17S)-3,3-dimethoxy-13-methyl-2,3,4,6,7,8,12,13,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-16,16,17-d3-17-ol
Molecular Weight (g/mol) 321.47
Formula Weight 321.24
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.