missing translation for 'onlineSavingsMsg'
Learn More

17alphabeta-Hydroxy-D-homo Betamethasone Ammonium Phosphate, TRC

Product Code. 30392746
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392746 5 mg 5mg
30395258 1 mg -
30406699 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392746 Supplier Toronto Research Chemicals Supplier No. TRCH9427755MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17alphabeta-Hydroxy-D-homo Betamethasone Ammonium Phosphate
Analyte or Component Names 17alphabeta-Hydroxy-D-homo Betamethasone Ammonium Phosphate
Shipping Condition Room Temperature
Molecular Formula C22 H29 F O8 P . H4 N
Synonym (1S,3S,4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-1,3,4,4a,4b,5,6,10a,10b,11,12,12a-dodecahydro-1,11-dihydroxy-3,10a,12a-trimethyl-1-[(phosphonooxy)methyl]-2,8-chrysenedione Ammonium Salt, 9α-Fluoro-11β,17α-dihydroxy-17α-[(phosphonooxy)methyl]-16β-methyl-D-homoandrosta-1,4-diene-3,17-dione Ammonium Salt
InChI Formula InChI=1S/C22H30FO8P.H3N/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)22(15,23)17(25)10-20(16,3)21(27,18(12)26)11-31-32(28,29)30;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);1H3/t12-,15-,16-,17-,19-,20-,21+,22-;/m0./s1
Solution Type Neat
SMILES [NH4+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@](O)(COP(=O)(O)[O-])C1=O
IUPAC Name azanium;[(1R,3S,4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-1,11-dihydroxy-3,10a,12a-trimethyl-2,8-dioxo-3,4,4a,4b,5,6,11,12-octahydrochrysen-1-yl]methyl hydrogen phosphate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 489.471
Quantity 5 mg
Formula Weight 489.193
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.