missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17alphabeta-Hydroxy-D-homo Betamethasone Ammonium Phosphate, TRC
Specifications
Specifications
| Chemical Name or Material | 17alphabeta-Hydroxy-D-homo Betamethasone Ammonium Phosphate |
| Analyte or Component Names | 17alphabeta-Hydroxy-D-homo Betamethasone Ammonium Phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H29 F O8 P . H4 N |
| Synonym | (1S,3S,4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-1,3,4,4a,4b,5,6,10a,10b,11,12,12a-dodecahydro-1,11-dihydroxy-3,10a,12a-trimethyl-1-[(phosphonooxy)methyl]-2,8-chrysenedione Ammonium Salt, 9α-Fluoro-11β,17α-dihydroxy-17α-[(phosphonooxy)methyl]-16β-methyl-D-homoandrosta-1,4-diene-3,17-dione Ammonium Salt |
| InChI Formula | InChI=1S/C22H30FO8P.H3N/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)22(15,23)17(25)10-20(16,3)21(27,18(12)26)11-31-32(28,29)30;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);1H3/t12-,15-,16-,17-,19-,20-,21+,22-;/m0./s1 |
| Solution Type | Neat |
| SMILES | [NH4+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@](O)(COP(=O)(O)[O-])C1=O |
| IUPAC Name | azanium;[(1R,3S,4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-1,11-dihydroxy-3,10a,12a-trimethyl-2,8-dioxo-3,4,4a,4b,5,6,11,12-octahydrochrysen-1-yl]methyl hydrogen phosphate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?