missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17Alpha-Trenbolone-d5 (Major), TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | 17alpha-Trenbolone-d5 |
| Molecular Formula | C18H17D5O2 |
| Quantity | 5 mg |
| Synonym | (17α)-17-Hydroxy-estra-4,9,11-trien-3-one-d5, 17-Epitrenbolone-d5, Epitrenbolone-d5, α-Trenbolone-d5 |
| InChI Formula | InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h8-10,15-17,20H,2-7H2,1H3/t15-,16+,17-,18+/m1/s1/i2D2,3D2,10D |
| SMILES | O[C@H]1[C@@]2(C)C=CC3=C4C(C([2H])([2H])C[C@@]3([H])[C@]2([H])CC1)=C([2H])C(C([2H])([2H])C4)=O |
| Molecular Weight (g/mol) | 275.4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction