missing translation for 'onlineSavingsMsg'
Learn More

17-Iodo-3-O-methyl Estratetraenol-d5, TRC

Product Code. 30404628
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404628 1 mg 1mg
30399797 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30404628 Supplier Toronto Research Chemicals Supplier No. TRCI7084221MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17-Iodo-3-O-methyl Estratetraenol-d5
Molecular Formula C19 D5 H18 I O
Quantity 1 mg
Synonym 17-Iodo-3-methoxy-estra-1,3,5(10),16-tetraene-d5
InChI Formula InChI=1S/C19H23IO/c1-19-10-9-15-14-6-4-13(21-2)11-12(14)3-5-16(15)17(19)7-8-18(19)20/h4,6,8,11,15-17H,3,5,7,9-10H2,1-2H3/t15-,16-,17+,19+/m1/s1/i3D2,5D2,15D
SMILES [2H]C1([2H])[C@H]2[C@@H]3CC=C(I)[C@@]3(C)CC[C@]2([2H])c4ccc(OC)cc4C1([2H])[2H]
IUPAC Name (8R,9S,13S,14S)-6,6,7,7,9-pentadeuterio-17-iodo-3-methoxy-13-methyl-11,12,14,15-tetrahydro-8H-cyclopenta[a]phenanthrene
Molecular Weight (g/mol) 399.321
Formula Weight 399.111
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.