missing translation for 'onlineSavingsMsg'
Learn More

17-Hydroxy-5Beta-pregnane-3,11,20-trione, TRC

Product Code. 30390675
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390675 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390675 Supplier Toronto Research Chemicals Supplier No. TRCH952260100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17-Hydroxy-5beta-pregnane-3,11,20-trione
CAS 10455-93-3
Molecular Formula C21H30O4
Quantity 100 mg
Synonym NSC 15470
InChI Formula InChI=1S/C21H30O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h13,15-16,18,25H,4-11H2,1-3H3/t13?,15-,16?,18?,19-,20-,21-/m0/s1
SMILES CC(=O)[C@@]1(O)CCC2[C@@H]3CCC4CC(=O)CC[C@]4(C)C3C(=O)C[C@]12C
IUPAC Name (8S,10S,13S,17R)-17-acetyl-17-hydroxy-10,13-dimethyl-1,2,4,5,6,7,8,9,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,11-dione
Molecular Weight (g/mol) 346.46
Formula Weight 346.2144
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.