missing translation for 'onlineSavingsMsg'
Learn More

17-Deshydroxyacetyl 17-Carbonyl Prednisolone, TRC

Product Code. 30388309
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388309 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388309 Supplier Toronto Research Chemicals Supplier No. TRCD679563100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17-Deshydroxyacetyl 17-Carbonyl Prednisolone
CAS 167997-18-4
Molecular Formula C20 H26 O4
Quantity 100 mg
Synonym (8S,9S,10R,11S,13S,14S,17R)-11,17-Dihydroxy-10,13-dimethyl-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-17-carbaldehyde
InChI Formula InChI=1S/C20H26O4/c1-18-7-5-13(22)9-12(18)3-4-14-15-6-8-20(24,11-21)19(15,2)10-16(23)17(14)18/h5,7,9,11,14-17,23-24H,3-4,6,8,10H2,1-2H3/t14-,15-,16-,17+,18-,19-,20-/m0/s1
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C=O
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-17-carbaldehyde
Molecular Weight (g/mol) 330.42
Formula Weight 330.1831
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.