missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17 beta-HSD14/HSD17B14 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-96996JF549
This item is not returnable.
View return policy
Description
17 beta-HSD14/HSD17B14 Polyclonal antibody specifically detects 17 beta-HSD14/HSD17B14 in Human samples. It is validated for ELISA
Specifications
| 17 beta-HSD14/HSD17B14 | |
| Polyclonal | |
| ELISA | |
| dehydrogenase/reductase (SDR family) member 10, Dehydrogenase/reductase SDR family member 10, DHRS1017-beta-hydroxysteroid dehydrogenase 14, EC 1.1.1.-, hydroxysteroid (17-beta) dehydrogenase 14,17-beta-hydroxysteroid dehydrogenase DHRS10, retinal short-chain dehydrogenase/reductase 3, Retinal short-chain dehydrogenase/reductase retSDR3, retSDR3, RETSDR3,17-beta-HSD 14, SDR3, SDR47C1, short chain dehydrogenase/reductase family 47C, member 1 | |
| Produced in rabbits immunized with purified, recombinant Human 17 beta-HSD14/HSD17B14 (Accession#: NP_057330.2; Met1-Ser270). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 51171 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction