missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17 beta-HSD14/HSD17B14 Antibody (6C4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42967
This item is not returnable.
View return policy
Description
17 beta-HSD14/HSD17B14 Monoclonal antibody specifically detects 17 beta-HSD14/HSD17B14 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| 17 beta-HSD14/HSD17B14 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| 6C4 | |
| Western Blot 1:1000 - 1:4000, ELISA | |
| dehydrogenase/reductase (SDR family) member 10, Dehydrogenase/reductase SDR family member 10, DHRS1017-beta-hydroxysteroid dehydrogenase 14, EC 1.1.1.-, hydroxysteroid (17-beta) dehydrogenase 14,17-beta-hydroxysteroid dehydrogenase DHRS10, retinal short-chain dehydrogenase/reductase 3, Retinal short-chain dehydrogenase/reductase retSDR3, retSDR3, RETSDR3,17-beta-HSD 14, SDR3, SDR47C1, short chain dehydrogenase/reductase family 47C, member 1 | |
| hydroxysteroid(17-beta) dehydrogenase 10 - (Uniprot# Q99714 ) | |
| 100 μg | |
| Lipid and Metabolism | |
| 51171 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction