missing translation for 'onlineSavingsMsg'
Learn More

17 beta-Estradiol, Lipomed

Product Code. 30462955
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462955 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462955 Supplier Lipomed Supplier No. LPMEST168450

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

17 beta-Estradiol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Estradiol
CAS 50-28-2
Analyte or Component Names Estradiol
Shipping Condition Room Temperature
Molecular Formula C18 H24 O2
Synonym Estra-1,3,5(10)-triene-3,17-diol (17β)-, Estradiol (8 CI), (17β)-Estra-1,3,5(10)-triene-3,17-diol, (+)-3,17β-Estradiol, 13β-Methyl-1,3,5(10)-gonatriene-3,17β-ol, 17β-Estradiol, 17β-Oestradiol, 3,17-Epidihydroxyestratriene, 3,17β-Dihydroxyestra-1,3,5(10)-triene, 3,17β-Estradiol, Absorlent Matrix, Aerodiol, Alora, Altrad, Aquadiol, Bardiol, Benzogynestry, Beta-estradiol, Bio-E Gel, Climaderm, Climara, Compudose, Compudose 100, Compudose 200, Compudose 365, Compudose 400, Corpagen, Dermestril, Destradiol, Dihydrofollicular hormone, Dihydrofolliculin, Dihydromenformon, Dihydrotheelin, Dihydroxyestrin, Dimenformon, Diogyn, Diogynets, Divigel, E 2, Elestrim, Encore, Epiestriol 50, Estasorb, Estra-1,3,5(10)-triene-3,17-diol, (17β)-, Estra-1,3,5(10)-triene-3,17β-diol, Estrace, Estraderm, Estraderm MX, Estraderm TTS, Estraderm TTS 100, Estraderm TTS 50, Estradiol-17β, Estradot, Estraldine, Estrapatch, Estredox, Estreva, Estring, Estring Vaginal Ring, Estroclim, Estroclim 50, Estrogel, Estrogel HBF, Estropause, Estrovite, Evamist, Evorel, FemSeven, Femanest, Femestral, Femogen, Fempatch, Follicyclin, Gelestra, Ginosedol, Gynergon, Gynodiol, Gynoestryl, Innofem, Lamdiol, Macrodiol, Menorest, Menostar, NSC 9895, Nordicol, Oesclim, Oestergon, Oestra-1,3,5(10)-triene-3,17β-diol, Oestradiol, Oestrogel, Oestroglandol, Ovahormon, Ovasterol, Ovastevol, Ovocyclin, Ovocylin, Perlatanol, Primofol, Profoliol, Profoliol B, Progynon, Progynon DH, Sandrena 1, Sprediol, Syndiol, Systen, Theelin, dihydro-, Vagifem, Vivelle, Vivelle-Dot, Zesteem, Zumenon, β-Estradiol
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES C[C@]12 CC[C@H]3 [C@@H](CCc4cc(O)ccc34)[C@@H]1 CC[C@@H]2 O
IUPAC Name (8 R,9 S,13 S,14 S,17 S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 272.38
Quantity 50 mg
Formula Weight 272.1776 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.