missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17-Beta-Estradiol Antibody (4S24 (BGN/06/8824)), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62280JF646
This item is not returnable.
View return policy
Description
17-Beta-Estradiol Monoclonal antibody specifically detects 17-Beta-Estradiol in All samples. It is validated for ELISA
Specifications
| 17-Beta-Estradiol | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 17 beta Oestradiol, 17-Beta-Estradiol, 17-beta-HSD | |
| 17-beta estradiol conjugated to bovine serum albumin at position 6. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| 4S24 (BGN/06/8824) | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction