missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17-Beta-Estradiol Antibody (4S24 (BGN/06/8824)), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62280AF750
This item is not returnable.
View return policy
Description
17-Beta-Estradiol Monoclonal specifically detects 17-Beta-Estradiol in All Species samples. It is validated for ELISA.
Specifications
| 17-Beta-Estradiol | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| NB100-62280 recognizes 17-beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. The following cross reactivites have been observed: 17-beta Estradiol 100% 17-beta Estradiol-BSA 90% Estriol <0.9% Testosterone <0.01% | |
| Store at 4°C in the dark. |
| ELISA | |
| 4S24 (BGN/06/8824) | |
| ELISA | |
| 17 beta Oestradiol, 17-Beta-Estradiol, 17-beta-HSD | |
| 17-beta estradiol conjugated to bovine serum albumin at position 6. | |
| Primary | |
| All species | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido