missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17,21-Dihydroxy-16b-methyl-pregna-1,4,9(11)-triene-3,20-dione 21-phosphate Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 17,21-Dihydroxy-16beta-methyl-pregna-1,4,9(11)-triene-3,20-dione 21-phosphate Ester |
| Analyte or Component Names | 17,21-Dihydroxy-16beta-methyl-pregna-1,4,9(11)-triene-3,20-dione 21-phosphate Ester |
| Shipping Condition | Blue Ice |
| Molecular Formula | C22H29O7P • 2Na |
| Synonym | 1-Hydroxy-2-((8S,10S,13S,14S,17R)-17-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,10,12,13,14,15,16,17-decahydro-3H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl dihydrogen Phosphate |
| InChI Formula | InChI=1S/C22H29O7P.2Na/c1-13-10-18-16-5-4-14-11-15(23)6-8-20(14,2)17(16)7-9-21(18,3)22(13,25)19(24)12-29-30(26,27)28;;/h6-8,11,13,16,18,25H,4-5,9-10,12H2,1-3H3,(H2,26,27,28);;/t13-,16+,18-,20-,21-,22-;;/m0../s1 |
| Solution Type | Neat |
| SMILES | O=C1C=C[C@@]2(C3=CC[C@@]4([C@](O)([C@H](C[C@]4([C@@]3(CCC2=C1)[H])[H])C)C(COP(O)(O)=O)=O)C)C.[Na].[Na] |
| IUPAC Name | 2-((8S,10S,13S,14S,16S,17R)-17-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,10,12,13,14,15,16,17-decahydro-3H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl dihydrogen phosphate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?