missing translation for 'onlineSavingsMsg'
Learn More

17,17-(Ethylenedioxy)androst-4-en-3Beta-ol, TRC

Product Code. 30391028
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391028 100 mg 100mg
30406034 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391028 Supplier Toronto Research Chemicals Supplier No. TRCE979660100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17,17-(Ethylenedioxy)androst-4-en-3beta-ol
CAS 2428-57-1
Molecular Formula C21H32O3
Quantity 100 mg
Synonym 3beta-Hydroxyandrost-4-en-17-one Cyclic Ethylene Acetal, Spiro[17H-cyclopenta[a]phenanthrene-17,2'-[1,3]dioxolane] Androst-4-en-17-one Deriv., (3beta)-3-Hydroxyandrost-4-en-17-one Cyclic 1,2-Ethanediyl Acetal
InChI Formula InChI=1S/C21H32O3/c1-19-8-5-15(22)13-14(19)3-4-16-17(19)6-9-20(2)18(16)7-10-21(20)23-11-12-24-21/h13,15-18,22H,3-12H2,1-2H3/t15-,16+,17-,18-,19-,20-/m0/s1
SMILES C[C@]12CC[C@H](O)C=C1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CCC45OCCO5
IUPAC Name (3S,8R,9S,10R,13S,14S)-10,13-dimethylspiro[1,2,3,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-17,2'-1,3-dioxolane]-3-ol
Molecular Weight (g/mol) 332.48
Formula Weight 332.2351
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.