missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrimeView™ 16 Global Gene Expression Profile Assay

Product Code. 18725535 Shop All Applied Biosystems Products
Click to view available options
Quantity:
16 assays
Unit Size:
16 tests
This item is not returnable. View return policy

Product Code. 18725535

Brand: Applied Biosystems™ 905402

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PrimeView 16 Global Gene Expression Profile Assay enables expression profiling of 16 samples at a time with an emphasis on established, well-annotated content.

The PrimeView 16 Global Gene Expression Profile Assay enables expression profiling of 16 samples at a time with an emphasis on established, well-annotated content. More than 36,000 transcripts and variants are measured per sample. Sequences used in the design of the array were selected from the UniGene database 219, RefSeq version 36, and full-length human mRNAs from GenBank™.

Note: The PluriTest™ Assay now accepts PrimeView array data from GeneTitan instruments.

Features include:

  • Cost-effective global confirmation of pluripotency marker expression
  • Simple-to-use online analysis tool
  • Published method with over 15,000 samples analyzed
  • For use with GeneTitan family of instruments

About the PluriTest Assay

The PluriTest Assay is a bioinformatics assay that compares the transcriptional profile of a sample to an extensive reference set of >450 cell/tissue types, including 223 hESC lines, 41 iPSC lines, somatic cells, and tissues. The online analysis tool confirms pluripotency marker expression via two separate scores: Pluripotency and Novelty. The Pluripotency Score informs the user on how strongly a model-based pluripotency signature is expressed in the samples analyzed. The Novelty Score indicates the general model fit for a given sample.

TRUSTED_SUSTAINABILITY

Specifications

Array Transcriptome Profiling
Content And Storage

• Arrays (store at 2°C to 8°C)
• Purification beads, Module 2 (2°C to 8°C)
• Module 1 (-5°C to -30°C)
• PolyA kit (-5°C to -30°C)
• Hybridization kit (-5°C to -30°C)
• Fragmention buffer, Module 2 (room temperature)

Format Assay
Product Line GeneChip, PrimeView
Product Type Gene Expression Profile Assay
Quantity 16 assays
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Species Human
For Use With (Application) Stem Cell Analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.