missing translation for 'onlineSavingsMsg'
Learn More

16Alpha-Hydroxycortisol, TRC

Product Code. 30400611
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400611 10 mg 10mg
30390465 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400611 Supplier Toronto Research Chemicals Supplier No. TRCH92228010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 16alpha-Hydroxycortisol
CAS 1171-81-9
Molecular Formula C21H30O6
Quantity 10 mg
Synonym U 34908, 16α,17-Dihydroxy-corticosterone, 11β,16α,17,21-Tetrahydroxy-pregn-4-ene-3,20-dione, 11β,16α,17,21-Tetrahydroxypregn-4-ene-3,20-dione
InChI Formula InChI=1S/C21H30O6/c1-19-6-5-12(23)7-11(19)3-4-13-14-8-16(25)21(27,17(26)10-22)20(14,2)9-15(24)18(13)19/h7,13-16,18,22,24-25,27H,3-6,8-10H2,1-2H3/t13-,14-,15-,16+,18+,19-,20-,21-/m0/s1
SMILES Cc1c2c(cccc2)nc1
Molecular Weight (g/mol) 378.46
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.