missing translation for 'onlineSavingsMsg'
Learn More
Learn More
16α-Homo Dexamethasone Phosphate, TRC
Specifications
Specifications
| Chemical Name or Material | 16Alpha-Homo Dexamethasone Phosphate |
| Analyte or Component Names | 16Alpha-Homo Dexamethasone Phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22H30FO8P |
| Synonym | ((4aS,4bS,10aS,10bR,11S,12aS)-10b-Fluoro-1,11-dihydroxy-3,10a,12a-trimethyl-2,8-dioxo-1,2,3,4,4a,4b,5,6,8,10a,10b,11,12,12a-tetradecahydrochrysen-1-yl)methyl Dihydrogen Phosphate, Dexamethasone Phosphate Impurity |
| InChI Formula | InChI=1S/C22H30FO8P/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)22(15,23)17(25)10-20(16,3)21(27,18(12)26)11-31-32(28,29)30/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30)/t12?,15-,16-,17-,19-,20-,21?,22-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12C(CC[C@@]3([C@@]1([C@]([H])(C[C@]4([C@]3(CC(C(C4(COP(O)(O)=O)O)=O)C)[H])C)O)F)[H])=CC(C=C2)=O |
| IUPAC Name | ((4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-1,11-dihydroxy-3,10a,12a-trimethyl-2,8-dioxo-1,2,3,4,4a,4b,5,6,8,10a,10b,11,12,12a-tetradecahydrochrysen-1-yl)methyl dihydrogen phosphate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?