missing translation for 'onlineSavingsMsg'
Learn More

16-epi Mometasone Furoate, TRC

Product Code. 30381815
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381815 10 mg -
30377924 25 mg -
30376832 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381815 Supplier Toronto Research Chemicals Supplier No. TRCD47468810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2231764-75-1
Molecular Formula C27H30Cl2O6
Molecular Weight (g/mol) 521.43
Synonym Pregna-1,4-diene-3,20-dione, 9,21-dichloro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-, (11β,16β)-, 9,21-Dichloro-11β-hydroxy-16β-methyl-3,20-dioxopregna-1,4-dien-17-yl Furan-2-carboxylate, 16-epi-Mometasone Furoate, Mometasone Furoate Imp. S (EP)
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-17-(2-chloroacetyl)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)CCl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H30Cl2O6/c1-15-11-19-18-7-6-16-12-17(30)8-9-24(16,2)26(18,29)21(31)13-25(19,3)27(15,22(32)14-28)35-23(33)20-5-4-10-34-20/h4-5,8-10,12,15,18-19,21,31H,6-7,11,13-14H2,1-3H3/t15-,18-,19-,21-,24-,25-,26-,27-/m0/s1
Formula Weight 520.1419
Chemical Name or Material 16-epi Mometasone Furoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.