missing translation for 'onlineSavingsMsg'
Learn More

16-b-Homo Betamethasone Phosphate, TRC

Product Code. 30389579
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389579 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389579 Supplier Toronto Research Chemicals Supplier No. TRCH1546201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 16-b-Homo Betamethasone Phosphate
CAS 1202002-02-5
Molecular Formula C22H30FO8P
Quantity 1 mg
Synonym (2S,3S,4aS,4bS,10aS,10bR,11S,12aS)-10b-Fluoro-3,4,4a,5,6,10a,10b,11,12,12a-decahydro-2,11-dihydroxy-3,10a,12a-trimethyl-2-[(phosphonooxy)methyl]-1,8(2H,4bH)-chrysenedione, ((2S,3S,4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-2,11-dihydroxy-3,10a,12a-trimethyl-1,8-dioxo-1,2,3,4,4a,4b,5,6,8,10a,10b,11,12,12a-tetradecahydrochrysen-2-yl)methyl dihydrogen phosphate, betamethasone impurity 86
InChI Formula InChI=1S/C22H30FO8P/c1-12-8-16-15-5-4-13-9-14(24)6-7-20(13,3)22(15,23)17(25)10-19(16,2)18(26)21(12,27)11-31-32(28,29)30/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30)/t12-,15-,16-,17-,19-,20-,21+,22-/m0/s1
SMILES F[C@@]12[C@]([C@]3([C@](C)(C[C@@H]1O)C(=O)[C@](COP(=O)(O)O)(O)[C@@H](C)C3)[H])(CCC=4[C@]2(C)C=CC(=O)C4)[H]
IUPAC Name ((2S,3S,4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-2,11-dihydroxy-3,10a,12a-trimethyl-1,8-dioxo-1,2,3,4,4a,4b,5,6,8,10a,10b,11,12,12a-tetradecahydrochrysen-2-yl)methyl dihydrogen phosphate
Molecular Weight (g/mol) 472.44
Formula Weight 472.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.