missing translation for 'onlineSavingsMsg'
Learn More

16,17Alpha-Epoxypregnenolone, TRC

Product Code. 30382887
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382887 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382887 Supplier Toronto Research Chemicals Supplier No. TRCE592315500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 16,17alpha-Epoxypregnenolone
CAS 974-23-2
Molecular Formula C21H30O3
Quantity 500 mg
Synonym 16alpha,17-Epoxy-3beta-hydroxypregn-5-en-20-one, 16alpha,17alpha-Epoxy-3beta-hydroxy-5-pregnen-20-one, 16alpha,17alpha-Epoxypregn-5-en-3beta-ol-20-one, 16alpha,17alpha-Epoxypregnenolone, 16alpha,17alpha-Oxidopregnenolone, 3beta-Hydroxy-16alpha,17alpha-epoxy-5-pregnen-20-one, 5-Pregnen-16alpha,17alpha-epoxy-3beta-ol-20-one, NSC 122401
InChI Formula InChI=1S/C21H30O3/c1-12(22)21-18(24-21)11-17-15-5-4-13-10-14(23)6-8-19(13,2)16(15)7-9-20(17,21)3/h4,14-18,23H,5-11H2,1-3H3/t14-,15+,16-,17-,18+,19-,20-,21+/m0/s1
SMILES CC(=O)[C@@]12O[C@@H]1C[C@H]3[C@@H]4CC=C5C[C@@H](O)CC[C@]5(C)[C@H]4CC[C@]23C
Molecular Weight (g/mol) 330.46
Formula Weight 330.2195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.