missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer PFA Tubing

Product Code. 30054248 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/2 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
7/16 in.
Diameter (English) Outer:
1/2 in.
1/4 in.
1/8 in.
3/16 in.
5/32 in.
9/16 in.
Unit Size:
15m
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Diameter (English) Outer unitSize
30054248 1/8 in. 3/16 in. 15m
30054049 1/2 in. 9/16 in. 15m
30054198 1/8 in. 1/4 in. 15m
30052975 7/16 in. 1/2 in. 15m
30053515 3/32 in. 5/32 in. 15m
30054256 3/16 in. 1/4 in. 15m
30054070 1/16 in. 1/8 in. 15m
30053735 5/32 in. 1/4 in. 15m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 30054248 Supplier Cole-Parmer Supplier No. 5011937
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Superior micro-porosity characteristics and e in., ternally smooth finishes—ideal for high-purity applications

Made from 100% virgin ultra-high molecular weight perfluoroalko in., y. E in., cellent fle in., ibility compared to other PFA grade tubing. Wide temperature range of -160 to 260 °C (-320 to 500 °F). Does not contain BPA, phthalates, or PVC. Sterilize by autoclave, EtO, and chemical sterilants. Complies with FDA 21 CFR 177.1550, USP Class VI, ASTM D 6867-03. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection

Specifications

Certifications/Compliance ADCF;FDA;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/8 in.
Diameter (English) Outer 3/16 in.
Overall Length (English) 50 ft.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.