missing translation for 'onlineSavingsMsg'
Learn More

15b-OH Gibberellin A3 Methyl Ester, TRC

Product Code. 30404692
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404692 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404692 Supplier Toronto Research Chemicals Supplier No. TRCG3774452.5MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 15beta-OH Gibberellin A3 Methyl Ester
CAS 105593-24-6
Molecular Formula C20H24O7
Quantity 2.5 mg
Synonym (1alpha,2beta,4aalpha,4bbeta,9beta,10beta)-2,4a,7,9-Tetrahydroxy-1-methyl-8-methylene-gibb-3-ene-1,10-dicarboxylic Acid 1,4a-Lactone 10-Methyl Ester, 4a,1-(Epoxymethano)-7,9a-methanobenz[a]azulene Gibb-3-ene-1,10-dicarboxylic Acid Derivative, (1S,2S,4aR,4bR,7S,9aR,10S)-Methyl 2,7,9-Trihydroxy-1-methyl-8-methylene-13-oxo-1,2,4b,5,6,7,8,9,10,10a-decahydro-4a,1-(epoxymethano)-7,9a-methanobenzo[a]azulene-10-carboxylate
InChI Formula InChI=1S/C20H24O7/c1-9-14(22)19-8-18(9,25)6-4-10(19)20-7-5-11(21)17(2,16(24)27-20)13(20)12(19)15(23)26-3/h5,7,10-14,21-22,25H,1,4,6,8H2,2-3H3/t10-,11+,12-,13?,14?,17-,18+,19-,20-/m1/s1
SMILES COC(=O)[C@H]1C2[C@@]3(C)[C@@H](O)C=C[C@@]2(OC3=O)[C@@H]4CC[C@]5(O)C[C@]14C(O)C5=C
Molecular Weight (g/mol) 376.4
Formula Weight 376.1522
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.