missing translation for 'onlineSavingsMsg'
Learn More

15b-OH Gibberellin A3 (>85%), TRC

Product Code. 30401629
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401629 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401629 Supplier Toronto Research Chemicals Supplier No. TRCG3774851MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 15beta-OH Gibberellin A3
CAS 105593-18-8
Molecular Formula C19H22O7
Quantity 1 mg
Synonym (1alpha,2beta,4aalpha,4bbeta,9beta,10beta)-2,4a,7,9-Tetrahydroxy-1-methyl-8-methylene-gibb-3-ene-1,10-dicarboxylic Acid 1,4a-Lactone
InChI Formula InChI=1S/C19H22O7/c1-8-13(21)18-7-17(8,25)5-3-9(18)19-6-4-10(20)16(2,15(24)26-19)12(19)11(18)14(22)23/h4,6,9-13,20-21,25H,1,3,5,7H2,2H3,(H,22,23)/t9-,10+,11-,12?,13?,16?,17+,18-,19-/m1/s1
SMILES CC12[C@@H](O)C=C[C@@]3(OC1=O)[C@@H]4CC[C@]5(O)C[C@]4(C(O)C5=C)[C@H](C23)C(=O)O
Molecular Weight (g/mol) 362.37
Formula Weight 362.1366
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.